Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGACACTAGCCCCCCATGCCTG[C/G]GTGGGAACAAGAAAACTCTAGGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613181 MIM: 185860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOLA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BOLA1 - bolA family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321025.1 | 523 | Missense Mutation | CGT,GGT | R,G 128 | NP_001307954.1 | |
NM_001321026.1 | 523 | Missense Mutation | CGT,GGT | R,G 128 | NP_001307955.1 | |
NM_016074.4 | 523 | Missense Mutation | CGT,GGT | R,G 128 | NP_057158.1 | |
XM_006711348.3 | 523 | Missense Mutation | CGT,GGT | R,G 128 | XP_006711411.1 |
SV2A - synaptic vesicle glycoprotein 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |