Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTGCCCCCCTACCCTCACCTG[A/G]GTACCAGCCCAGGGGCCTCGGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBQLN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBQLN4 - ubiquilin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304342.1 | Intron | NP_001291271.1 | ||||
NM_020131.4 | Intron | NP_064516.2 | ||||
XM_005245348.3 | Intron | XP_005245405.1 |