Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCTTCCTCGGAGGGGCCCCTC[A/G]AGACACTGGGCTGCGGGTGCCTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610595 MIM: 607377 MIM: 607646 | ||||||||||||||||||||
Literature Links: |
FLAD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLAD1 - flavin adenine dinucleotide synthetase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184891.1 | 84 | Intron | NP_001171820.1 | |||
NM_001184892.1 | 84 | Intron | NP_001171821.1 | |||
NM_025207.4 | 84 | Intron | NP_079483.3 | |||
NM_201398.2 | 84 | Intron | NP_958800.1 |
LENEP - lens epithelial protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018655.2 | 84 | Missense Mutation | CAA,CGA | Q,R 21 | NP_061125.1 |
ZBTB7B - zinc finger and BTB domain containing 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |