Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGCAGACAACCCCGAGAGTGTC[C/T]GCCACAGCCTCAGCCAGACATCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTPCR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NTPCR - nucleoside-triphosphatase, cancer-related | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCNX2 - pecanex homolog 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014801.3 | 6120 | Silent Mutation | GCA,GCG | A,A 2107 | NP_055616.3 | |
XM_006711816.3 | 6120 | Intron | XP_006711879.1 | |||
XM_011544278.2 | 6120 | Silent Mutation | GCA,GCG | A,A 1365 | XP_011542580.2 | |
XM_017002391.1 | 6120 | Silent Mutation | GCA,GCG | A,A 1904 | XP_016857880.1 | |
XM_017002392.1 | 6120 | Silent Mutation | GCA,GCG | A,A 961 | XP_016857881.1 |