Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTGCTGAGTGCTGATGATGCTC[C/T]GATGCCTGTCAAATTCCTAGCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607560 MIM: 609043 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF2 - Rho/Rac guanine nucleotide exchange factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIAA0907 - KIAA0907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6738 - microRNA 6738 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RXFP4 - relaxin/insulin like family peptide receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181885.2 | 110 | Missense Mutation | CCG,CTG | P,L 30 | NP_871001.1 |