Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCTGCCTGAGAACTGGACAGACA[C/T]GCGGGAGACGCTGCTGGAGGGGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605747 | ||||||||||||||||||||
Literature Links: |
LDLRAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LDLRAP1 - low density lipoprotein receptor adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015627.2 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | NP_056442.2 | |
XM_006710559.3 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_006710622.1 | |
XM_006710560.3 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_006710623.1 | |
XM_006710561.3 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_006710624.1 | |
XM_011541209.2 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_011539511.1 | |
XM_011541210.2 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_011539512.1 | |
XM_011541211.2 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_011539513.1 | |
XM_011541212.2 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_011539514.1 | |
XM_017000994.1 | 1169 | Missense Mutation | ACG,ATG | T,M 11 | XP_016856483.1 | |
XM_017000995.1 | 1169 | Missense Mutation | ACG,ATG | T,M 38 | XP_016856484.1 |