Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTAGTTTTCTTACCTTTATTTC[C/T]TTGACTGAAATGACAGAAAGCACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602105 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB17 - ankyrin repeat and SOCS box containing 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080868.2 | 812 | Silent Mutation | AAA,AAG | K,K 224 | NP_543144.1 | |
XM_006710345.3 | 812 | Silent Mutation | AAA,AAG | K,K 176 | XP_006710408.1 | |
XM_006710346.3 | 812 | Silent Mutation | AAA,AAG | K,K 146 | XP_006710409.1 |
MSH4 - mutS homolog 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |