Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCGCTCCAACACCAGGTCCGGG[A/G]AGATGCCACCTTCCAGGCTGGTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614269 MIM: 609971 | ||||||||||||||||||||
Literature Links: |
GPR153 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR153 - G protein-coupled receptor 153 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207370.2 | 1350 | Missense Mutation | TCC,TTC | S,F 335 | NP_997253.2 | |
XM_011541434.2 | 1350 | Missense Mutation | TCC,TTC | S,F 335 | XP_011539736.1 | |
XM_017001250.1 | 1350 | Missense Mutation | TCC,TTC | S,F 335 | XP_016856739.1 | |
XM_017001251.1 | 1350 | Missense Mutation | TCC,TTC | S,F 335 | XP_016856740.1 |
HES3 - hes family bHLH transcription factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |