Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCTGATGTCCAGCAATGTGGGC[A/G]GTGGCTTGTTCATCGGCCTGGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SLC5A9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC5A9 - solute carrier family 5 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011547.2 | 308 | Missense Mutation | AGT,GGT | S,G 90 | NP_001011547.2 | |
NM_001135181.1 | 308 | Missense Mutation | AGT,GGT | S,G 90 | NP_001128653.1 | |
XM_011540924.2 | 308 | Missense Mutation | AGT,GGT | S,G 90 | XP_011539226.1 | |
XM_011540925.2 | 308 | Missense Mutation | AGT,GGT | S,G 90 | XP_011539227.1 | |
XM_011540926.2 | 308 | UTR 5 | XP_011539228.1 | |||
XM_011540927.1 | 308 | UTR 5 | XP_011539229.1 | |||
XM_011540928.1 | 308 | UTR 5 | XP_011539230.1 | |||
XM_011540929.2 | 308 | Missense Mutation | AGT,GGT | S,G 90 | XP_011539231.1 | |
XM_017000558.1 | 308 | Missense Mutation | AGT,GGT | S,G 90 | XP_016856047.1 |