Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCAGGTTGCTTCGAGGAGACGCC[C/G]TGGTGGAGCTGGGCCTCAACGATTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605548 MIM: 601380 | ||||||||||||||||||||
Literature Links: |
ADAM15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM15 - ADAM metallopeptidase domain 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EFNA4 - ephrin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005227.2 | 220 | Missense Mutation | CTG,GTG | L,V 45 | NP_005218.1 | |
NM_182689.1 | 220 | Missense Mutation | CTG,GTG | L,V 45 | NP_872631.1 | |
NM_182690.2 | 220 | Missense Mutation | CTG,GTG | L,V 45 | NP_872632.2 |
LOC100505666 - uncharacterized LOC100505666 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |