Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCAGCTGCACCGGGCCCCCAGC[C/G]ATCCCTGGCATCCCGGGTATCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120570 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QB - complement C1q B chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000491.3 | 240 | Silent Mutation | GCC,GCG | A,A 36 | NP_000482.3 | |
XM_011542059.2 | 240 | Silent Mutation | GCC,GCG | A,A 36 | XP_011540361.1 |