Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCTGCTCCTGTTCCAACATCA[C/T]CTTCTCTCAGTTCCTCTGGTCGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600114 | ||||||||||||||||||||
Literature Links: |
CCT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCT3 - chaperonin containing TCP1 subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008800.2 | 1139 | Missense Mutation | GAT,GGT | D,G 303 | NP_001008800.1 | |
NM_005998.4 | 1139 | Missense Mutation | GAT,GGT | D,G 341 | NP_005989.3 |