Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGGGAGCCTTGGGAGTGCCAGC[C/T]GGGCTCTTGGTGGGAGGCTGAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR157 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR157 - G protein-coupled receptor 157 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024980.4 | 818 | Silent Mutation | CCA,CCG | P,P 307 | NP_079256.4 | |
XM_005263496.4 | 818 | Silent Mutation | CCA,CCG | P,P 294 | XP_005263553.1 | |
XM_005263497.4 | 818 | Silent Mutation | CCA,CCG | P,P 242 | XP_005263554.1 |