Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCACCGCTCACTGGAGAGGCCC[A/T]CTCTTTCCAGGCGTCTCTTCAGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605393 | ||||||||||||||||||||
Literature Links: |
KDM5B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KDM5B - lysine demethylase 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314042.1 | 5523 | Missense Mutation | GAG,GTG | E,V 1453 | NP_001300971.1 | |
NM_006618.4 | 5523 | Missense Mutation | GAG,GTG | E,V 1417 | NP_006609.3 | |
XM_011509088.2 | 5523 | Missense Mutation | GAG,GTG | E,V 1372 | XP_011507390.1 | |
XM_011509090.2 | 5523 | Missense Mutation | GAG,GTG | E,V 1295 | XP_011507392.1 | |
XM_011509091.2 | 5523 | Missense Mutation | GAG,GTG | E,V 1295 | XP_011507393.1 | |
XM_011509092.2 | 5523 | Missense Mutation | GAG,GTG | E,V 1259 | XP_011507394.1 |