Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGAGCGCTTGGATTTATGTATAT[A/T]AGGTGAGCGTACATTTATGTACATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
PRPF38B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRPF38B - pre-mRNA processing factor 38B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018061.2 | 795 | Silent Mutation | ATA,ATT | I,I 165 | NP_060531.2 | |
XM_011541687.2 | 795 | Silent Mutation | ATA,ATT | I,I 165 | XP_011539989.1 | |
XM_017001596.1 | 795 | Silent Mutation | ATA,ATT | I,I 20 | XP_016857085.1 | |
XM_017001597.1 | 795 | UTR 5 | XP_016857086.1 | |||
XM_017001598.1 | 795 | UTR 5 | XP_016857087.1 |