Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGAACTTCTCTATTTAAAAAAAA[A/G]GAGTAGGTCTAAAATTAAATTATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCARNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SCARNA2 - small Cajal body-specific RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM167B - transmembrane protein 167B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322248.1 | 1735 | UTR 3 | NP_001309177.1 | |||
NM_020141.3 | 1735 | UTR 3 | NP_064526.1 |