Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGGTCATCGCCCCGTATCTCCA[A/G]CGCCGGGAGCCTCGGGACTGCGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 180610 | ||||||||||||||||||||
Literature Links: |
RLF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RLF - rearranged L-myc fusion | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012421.3 | 158 | Missense Mutation | ACG,GCG | T,A 43 | NP_036553.2 | |
XM_017002016.1 | 158 | UTR 5 | XP_016857505.1 | |||
XM_017002017.1 | 158 | UTR 5 | XP_016857506.1 | |||
XM_017002018.1 | 158 | UTR 5 | XP_016857507.1 |