Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATAATTTACTACAGGTCTCTGA[A/C]GTGTATATAAAATGTTTTAGTGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 153420 MIM: 603491 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD58 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD58 - CD58 molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IGSF3 - immunoglobulin superfamily member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007237.2 | 7147 | UTR 3 | NP_001007238.1 | |||
NM_001542.3 | 7147 | UTR 3 | NP_001533.2 | |||
XM_005270793.2 | 7147 | UTR 3 | XP_005270850.1 | |||
XM_005270794.4 | 7147 | UTR 3 | XP_005270851.1 | |||
XM_006710593.3 | 7147 | UTR 3 | XP_006710656.1 | |||
XM_011541315.1 | 7147 | UTR 3 | XP_011539617.1 | |||
XM_011541316.2 | 7147 | UTR 3 | XP_011539618.1 |