Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCGAGGATTAGCGGGCTCTGAAC[A/G]CAGACTCCGCACTGACGGGACGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EPB41 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EPB41 - erythrocyte membrane protein band 4.1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM200B - transmembrane protein 200B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003682.3 | 693 | Missense Mutation | CGT,TGT | R,C 207 | NP_001003682.1 | |
NM_001171868.1 | 693 | Missense Mutation | CGT,TGT | R,C 207 | NP_001165339.1 | |
XM_011541457.2 | 693 | Missense Mutation | CGT,TGT | R,C 218 | XP_011539759.1 |