Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGTAATATTGCATGACTCACA[C/G]ACTCCTTGGACTTTAAATATTTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611467 | ||||||||||||||||||||
Literature Links: |
GBP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GBP5 - guanylate binding protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134486.2 | 1832 | Missense Mutation | TCT,TGT | S,C 468 | NP_001127958.1 | |
NM_052942.3 | 1832 | Missense Mutation | TCT,TGT | S,C 468 | NP_443174.1 |