Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACGAATGAGGGGACCACATCTA[C/T]GCTGACAATGAATCCTGTTAGTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 192225 | ||||||||||||||||||||
Literature Links: |
VCAM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VCAM1 - vascular cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001078.3 | 457 | Missense Mutation | ACG,ATG | T,M 79 | NP_001069.1 | |
NM_001199834.1 | 457 | Intron | NP_001186763.1 | |||
NM_080682.2 | 457 | Missense Mutation | ACG,ATG | T,M 79 | NP_542413.1 |