Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGACATGCTTTTGTTTCCCAC[C/T]GACTGTTTAATTTTTTTGGCTGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605960 MIM: 182891 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOSC10 - exosome component 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001998.2 | 2662 | Silent Mutation | TCA,TCG | S,S 861 | NP_001001998.1 | |
NM_002685.3 | 2662 | Silent Mutation | TCA,TCG | S,S 836 | NP_002676.1 | |
XM_005263475.2 | 2662 | Silent Mutation | TCA,TCG | S,S 866 | XP_005263532.2 | |
XM_017001494.1 | 2662 | Silent Mutation | TCA,TCG | S,S 450 | XP_016856983.1 | |
XM_017001495.1 | 2662 | Silent Mutation | TCA,TCG | S,S 445 | XP_016856984.1 | |
XM_017001496.1 | 2662 | Silent Mutation | TCA,TCG | S,S 434 | XP_016856985.1 | |
XM_017001497.1 | 2662 | Silent Mutation | TCA,TCG | S,S 429 | XP_016856986.1 |
SRM - spermidine synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |