Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATGAATGCCGCCCTGGTTATTC[C/T]GGAAGACCGTTTTCTATCATCTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CR1 - complement component 3b/4b receptor 1 (Knops blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000573.3 | 350 | Silent Mutation | TCC,TCT | S,S 78 | NP_000564.2 | |
NM_000651.4 | 350 | Silent Mutation | TCC,TCT | S,S 78 | NP_000642.3 | |
XM_006711166.2 | 350 | Silent Mutation | TCC,TCT | S,S 83 | XP_006711229.1 | |
XM_011509205.1 | 350 | Silent Mutation | TCC,TCT | S,S 83 | XP_011507507.1 |