Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAATTACCTTGTCGGTCCTGCTG[A/C]CACGGTTTGGGCGCCCACCACGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607378 | ||||||||||||||||||||
Literature Links: |
SERBP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERBP1 - SERPINE1 mRNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018067.1 | 1231 | Missense Mutation | GGC,GTC | G,V 385 | NP_001018077.1 | |
NM_001018068.1 | 1231 | Missense Mutation | GGC,GTC | G,V 379 | NP_001018078.1 | |
NM_001018069.1 | 1231 | Missense Mutation | GGC,GTC | G,V 370 | NP_001018079.1 | |
NM_015640.3 | 1231 | Missense Mutation | GGC,GTC | G,V 364 | NP_056455.3 |