Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCTGGCACTTGTGAGGCTTCTC[A/T]CCTGTGGGGATGGGAGGGGGAGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600871 | ||||||||||||||||||||
Literature Links: |
GFI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GFI1 - growth factor independent 1 transcriptional repressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127215.1 | 1460 | Silent Mutation | GGA,GGT | G,G 364 | NP_001120687.1 | |
NM_001127216.1 | 1460 | Silent Mutation | GGA,GGT | G,G 364 | NP_001120688.1 | |
NM_005263.3 | 1460 | Silent Mutation | GGA,GGT | G,G 364 | NP_005254.2 | |
XM_005270749.3 | 1460 | Silent Mutation | GGA,GGT | G,G 364 | XP_005270806.1 | |
XM_011541245.2 | 1460 | Silent Mutation | GGA,GGT | G,G 364 | XP_011539547.1 | |
XM_011541246.2 | 1460 | Silent Mutation | GGA,GGT | G,G 364 | XP_011539548.1 |