Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGACGAACTTCAGCACCTGCTCA[C/T]TGGACAGGCCCTGGTAGGGTTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147671 MIM: 191315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INSRR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INSRR - insulin receptor related receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014215.2 | 3892 | Missense Mutation | AAT,AGT | N,S 1198 | NP_055030.1 |
NTRK1 - neurotrophic receptor tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007792.1 | 3892 | Intron | NP_001007793.1 | |||
NM_001012331.1 | 3892 | Intron | NP_001012331.1 | |||
NM_002529.3 | 3892 | Intron | NP_002520.2 |