Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAACTGCAGCAGGGGCGCAGGAT[A/G]GACAAAGGACCAGCTCGGGCTCGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606373 | ||||||||||||||||||||
Literature Links: |
FMN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FMN2 - formin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305424.1 | 739 | Missense Mutation | AGA,GGA | R,G 172 | NP_001292353.1 | |
NM_020066.4 | 739 | Missense Mutation | AGA,GGA | R,G 172 | NP_064450.3 | |
XM_011544237.2 | 739 | Missense Mutation | AGA,GGA | R,G 172 | XP_011542539.1 | |
XM_017001837.1 | 739 | Missense Mutation | AGA,GGA | R,G 172 | XP_016857326.1 | |
XM_017001838.1 | 739 | Missense Mutation | AGA,GGA | R,G 172 | XP_016857327.1 | |
XM_017001839.1 | 739 | Intron | XP_016857328.1 | |||
XM_017001840.1 | 739 | Intron | XP_016857329.1 | |||
XM_017001841.1 | 739 | Intron | XP_016857330.1 | |||
XM_017001842.1 | 739 | Missense Mutation | AGA,GGA | R,G 172 | XP_016857331.1 | |
XM_017001843.1 | 739 | Missense Mutation | AGA,GGA | R,G 172 | XP_016857332.1 |