Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTCCCAGCTCTGGGGGCTGCTG[C/T]GGCTCCAGCTCTGGGGGCTGCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612609 MIM: 612618 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCE2A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCE2A - late cornified envelope 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178428.3 | 207 | Silent Mutation | TGC,TGT | C,C 52 | NP_848515.1 |
LCE4A - late cornified envelope 4A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |