Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGCTCACCTGCTCCTTGTTGT[G/T]CTCCAAAGCGGGCAGCACCTCTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 613466 | ||||||||||||||||||||
Literature Links: |
KLHDC9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHDC9 - kelch domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PFDN2 - prefoldin subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012394.3 | 337 | Missense Mutation | AAC,CAC | N,H 92 | NP_036526.2 | |
XM_011509624.2 | 337 | Missense Mutation | AAC,CAC | N,H 44 | XP_011507926.1 |