Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCGGCGGAGAGCAGCACCCAGC[A/G]GCGCCCCGGGGTCCCCGCGGGACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST6GALNAC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST6GALNAC5 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320273.1 | 465 | Missense Mutation | CAG,CGG | Q,R 66 | NP_001307202.1 | |
NM_001320274.1 | 465 | Missense Mutation | CAG,CGG | Q,R 66 | NP_001307203.1 | |
NM_030965.2 | 465 | Missense Mutation | CAG,CGG | Q,R 66 | NP_112227.1 |