Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAACTATTACCCATCTTTTAAAA[G/T]CAGGAGACCAAATTATTTGTATGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607657 | ||||||||||||||||||||
Literature Links: |
CTH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTH - cystathionine gamma-lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190463.1 | 505 | Intron | NP_001177392.1 | |||
NM_001902.5 | 505 | Missense Mutation | GCA,TCA | A,S 103 | NP_001893.2 | |
NM_153742.4 | 505 | Missense Mutation | GCA,TCA | A,S 103 | NP_714964.2 | |
XM_005270509.3 | 505 | UTR 5 | XP_005270566.1 | |||
XM_017000416.1 | 505 | Intron | XP_016855905.1 |