Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCCGGCGGTGAGGCGCTTCATG[A/G]AGAACTTCGGGGGAATCTCGTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 137290 | ||||||||||||||||||||
Literature Links: |
TACSTD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TACSTD2 - tumor-associated calcium signal transducer 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002353.2 | 1144 | Missense Mutation | TCC,TTC | S,F 269 | NP_002344.2 |