Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAACAATGGCTTACATTGATTACA[A/G]TATGATTGTAGCCTTTATGCTTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFI44L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFI44L - interferon induced protein 44 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006820.3 | 214 | Missense Mutation | AAT,AGT | N,S 59 | NP_006811.2 | |
XM_005270391.3 | 214 | Intron | XP_005270448.1 | |||
XM_005270392.4 | 214 | Intron | XP_005270449.1 | |||
XM_005270393.4 | 214 | Intron | XP_005270450.1 | |||
XM_006710304.3 | 214 | Intron | XP_006710367.1 | |||
XM_011540539.2 | 214 | Missense Mutation | AAT,AGT | N,S 59 | XP_011538841.1 | |
XM_017000120.1 | 214 | Intron | XP_016855609.1 |