Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGGGGTGGCTCCTACCCACACCTG[C/T]TGTGGGACGTGAGGAAAAGGTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610494 MIM: 614737 | ||||||||||||||||||||
Literature Links: |
DCAF6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DCAF6 - DDB1 and CUL4 associated factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017977.2 | 325 | Silent Mutation | CTG,TTG | L,L 11 | NP_001017977.1 | |
NM_001198956.1 | 325 | Silent Mutation | CTG,TTG | L,L 11 | NP_001185885.1 | |
NM_001198957.1 | 325 | Silent Mutation | CTG,TTG | L,L 11 | NP_001185886.1 | |
NM_018442.3 | 325 | Silent Mutation | CTG,TTG | L,L 11 | NP_060912.2 | |
XM_005245331.4 | 325 | Silent Mutation | CTG,TTG | L,L 11 | XP_005245388.1 | |
XM_005245332.4 | 325 | Silent Mutation | CTG,TTG | L,L 11 | XP_005245389.1 | |
XM_005245333.4 | 325 | Silent Mutation | CTG,TTG | L,L 11 | XP_005245390.1 | |
XM_017001779.1 | 325 | Intron | XP_016857268.1 | |||
XM_017001780.1 | 325 | Intron | XP_016857269.1 | |||
XM_017001781.1 | 325 | Intron | XP_016857270.1 |
MPC2 - mitochondrial pyruvate carrier 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143674.3 | 325 | UTR 5 | NP_001137146.1 | |||
NM_015415.3 | 325 | Intron | NP_056230.1 | |||
XM_006711266.3 | 325 | Intron | XP_006711329.1 |