Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGAATATGGCATAAGTATGGAG[C/T]CATACATCCTTACTGGAAATTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615100 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTTNBP2NL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTTNBP2NL - CTTNBP2 N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018704.2 | Intron | NP_061174.1 | ||||
XM_011541781.2 | Intron | XP_011540083.1 | ||||
XM_017001806.1 | Intron | XP_016857295.1 |