Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATATTTCATGCTTTTCAGTGGGCA[G/T]TGGGAATGAGAAGGACAGATGATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEK7 - NIMA related kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133494.2 | Intron | NP_598001.1 | ||||
XM_011509209.1 | Intron | XP_011507511.1 | ||||
XM_017000344.1 | Intron | XP_016855833.1 | ||||
XM_017000345.1 | Intron | XP_016855834.1 | ||||
XM_017000346.1 | Intron | XP_016855835.1 | ||||
XM_017000347.1 | Intron | XP_016855836.1 | ||||
XM_017000348.1 | Intron | XP_016855837.1 |