Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAATTATGGTTGGACTTCGTGCTT[A/G]TTGTACAGTCTTTGTGATAGACTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616119 MIM: 602413 | ||||||||||||||||||||
Literature Links: |
CFAP126 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CFAP126 - cilia and flagella associated protein 126 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013625.3 | 424 | Nonsense Mutation | CAA,TAA | Q,* 137 | NP_001013647.2 |
SDHC - succinate dehydrogenase complex subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035511.1 | 424 | Intron | NP_001030588.1 | |||
NM_001035512.1 | 424 | Intron | NP_001030589.1 | |||
NM_001035513.1 | 424 | Intron | NP_001030590.1 | |||
NM_001278172.1 | 424 | Intron | NP_001265101.1 | |||
NM_003001.3 | 424 | Intron | NP_002992.1 |