Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACTTTTCCGAAGCTCTCGGTC[C/T]TGGATGAATCCTGCAAACATCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605783 | ||||||||||||||||||||
Literature Links: |
BCAS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCAS2 - breast carcinoma amplified sequence 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DENND2C - DENN domain containing 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256404.1 | 3339 | Silent Mutation | CAA,CAG | Q,Q 879 | NP_001243333.1 | |
NM_198459.3 | 3339 | Silent Mutation | CAA,CAG | Q,Q 822 | NP_940861.3 |