Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTGGATCTTGATCTCGGGACA[C/T]GCAGGGTCCCCCAACTTGCCGGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAP1GAP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAP1GAP - RAP1 GTPase activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145657.1 | 2187 | Silent Mutation | GCA,GCG | A,A 661 | NP_001139129.1 | |
NM_001145658.1 | 2187 | Silent Mutation | GCA,GCG | A,A 707 | NP_001139130.1 | |
NM_002885.2 | 2187 | Silent Mutation | GCA,GCG | A,A 643 | NP_002876.2 | |
XM_005245955.3 | 2187 | Silent Mutation | GCA,GCG | A,A 661 | XP_005246012.2 | |
XM_006710804.2 | 2187 | Silent Mutation | GCA,GCG | A,A 728 | XP_006710867.2 | |
XM_006710805.2 | 2187 | Silent Mutation | GCA,GCG | A,A 720 | XP_006710868.2 | |
XM_017001965.1 | 2187 | Silent Mutation | GCA,GCG | A,A 728 | XP_016857454.1 | |
XM_017001966.1 | 2187 | Silent Mutation | GCA,GCG | A,A 720 | XP_016857455.1 | |
XM_017001967.1 | 2187 | Silent Mutation | GCA,GCG | A,A 690 | XP_016857456.1 | |
XM_017001968.1 | 2187 | Silent Mutation | GCA,GCG | A,A 669 | XP_016857457.1 | |
XM_017001969.1 | 2187 | Silent Mutation | GCA,GCG | A,A 669 | XP_016857458.1 | |
XM_017001970.1 | 2187 | Silent Mutation | GCA,GCG | A,A 661 | XP_016857459.1 | |
XM_017001971.1 | 2187 | Silent Mutation | GCA,GCG | A,A 784 | XP_016857460.1 | |
XM_017001972.1 | 2187 | Silent Mutation | GCA,GCG | A,A 635 | XP_016857461.1 | |
XM_017001973.1 | 2187 | Silent Mutation | GCA,GCG | A,A 643 | XP_016857462.1 | |
XM_017001974.1 | 2187 | Silent Mutation | GCA,GCG | A,A 631 | XP_016857463.1 | |
XM_017001975.1 | 2187 | Silent Mutation | GCA,GCG | A,A 635 | XP_016857464.1 | |
XM_017001976.1 | 2187 | Silent Mutation | GCA,GCG | A,A 605 | XP_016857465.1 | |
XM_017001977.1 | 2187 | Silent Mutation | GCA,GCG | A,A 597 | XP_016857466.1 | |
XM_017001978.1 | 2187 | Silent Mutation | GCA,GCG | A,A 728 | XP_016857467.1 | |
XM_017001979.1 | 2187 | Silent Mutation | GCA,GCG | A,A 720 | XP_016857468.1 | |
XM_017001980.1 | 2187 | Silent Mutation | GCA,GCG | A,A 699 | XP_016857469.1 | |
XM_017001981.1 | 2187 | Silent Mutation | GCA,GCG | A,A 669 | XP_016857470.1 | |
XM_017001982.1 | 2187 | Silent Mutation | GCA,GCG | A,A 669 | XP_016857471.1 | |
XM_017001983.1 | 2187 | Silent Mutation | GCA,GCG | A,A 669 | XP_016857472.1 | |
XM_017001984.1 | 2187 | Silent Mutation | GCA,GCG | A,A 661 | XP_016857473.1 | |
XM_017001985.1 | 2187 | Silent Mutation | GCA,GCG | A,A 661 | XP_016857474.1 | |
XM_017001986.1 | 2187 | Silent Mutation | GCA,GCG | A,A 643 | XP_016857475.1 | |
XM_017001987.1 | 2187 | Silent Mutation | GCA,GCG | A,A 643 | XP_016857476.1 | |
XM_017001988.1 | 2187 | Silent Mutation | GCA,GCG | A,A 643 | XP_016857477.1 | |
XM_017001989.1 | 2187 | Silent Mutation | GCA,GCG | A,A 635 | XP_016857478.1 | |
XM_017001990.1 | 2187 | Silent Mutation | GCA,GCG | A,A 635 | XP_016857479.1 | |
XM_017001991.1 | 2187 | Silent Mutation | GCA,GCG | A,A 605 | XP_016857480.1 |