Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCATCAGAGAGCCACAGTCCTAA[A/G]TAGACTCCTCGGTGCGCTCTGCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609318 MIM: 604718 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM45 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM45 - tripartite motif containing 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145635.1 | 2311 | Missense Mutation | NP_001139107.1 | |||
NM_025188.3 | 2311 | Missense Mutation | NP_079464.2 | |||
XM_011542199.2 | 2311 | Missense Mutation | XP_011540501.1 |
TTF2 - transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |