Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTGCAAGCTAGGTCGTCGACTG[C/T]AGGAAATCAGTACTACTTTAGGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616836 MIM: 611032 | ||||||||||||||||||||
Literature Links: |
GPATCH2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPATCH2 - G-patch domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297754.1 | 96 | Intron | NP_001284683.1 | |||
NM_018040.3 | 96 | Intron | NP_060510.1 | |||
XM_011509689.2 | 96 | Intron | XP_011507991.1 | |||
XM_011509690.2 | 96 | Intron | XP_011507992.1 | |||
XM_011509691.2 | 96 | Intron | XP_011507993.1 | |||
XM_011509693.2 | 96 | Intron | XP_011507995.1 | |||
XM_011509694.2 | 96 | Intron | XP_011507996.1 | |||
XM_017001592.1 | 96 | Intron | XP_016857081.1 | |||
XM_017001593.1 | 96 | Intron | XP_016857082.1 |
SPATA17 - spermatogenesis associated 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138796.3 | 96 | Missense Mutation | GCA,GTA | A,V 14 | NP_620151.1 | |
XM_005273052.1 | 96 | Missense Mutation | GCA,GTA | A,V 14 | XP_005273109.1 | |
XM_006711165.3 | 96 | Missense Mutation | GCA,GTA | A,V 14 | XP_006711228.1 | |
XM_011509194.2 | 96 | Missense Mutation | GCA,GTA | A,V 14 | XP_011507496.1 |