Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGACCATCTCTGGGACAGAGG[C/T]GACCAGGAAGCCAAGACCGGCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600426 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
E2F2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
E2F2 - E2F transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004091.3 | 1919 | UTR 3 | NP_004082.1 | |||
XM_005245748.3 | 1919 | UTR 3 | XP_005245805.1 | |||
XM_005245749.3 | 1919 | UTR 3 | XP_005245806.1 | |||
XM_011540868.2 | 1919 | UTR 3 | XP_011539170.1 | |||
XM_011540870.2 | 1919 | Intron | XP_011539172.1 | |||
XM_011540871.2 | 1919 | UTR 3 | XP_011539173.1 |