Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGATTTACAAGAGTCTTTAAAGCA[C/T]GGACTTACACCTATTGGTAAGTAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604453 | ||||||||||||||||||||
Literature Links: |
NR5A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NR5A2 - nuclear receptor subfamily 5 group A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276464.1 | 294 | Intron | NP_001263393.1 | |||
NM_003822.4 | 294 | Silent Mutation | CAC,CAT | H,H 16 | NP_003813.1 | |
NM_205860.2 | 294 | Silent Mutation | CAC,CAT | H,H 16 | NP_995582.1 | |
XM_005245062.3 | 294 | Intron | XP_005245119.1 | |||
XM_011509380.1 | 294 | UTR 5 | XP_011507682.1 | |||
XM_011509381.2 | 294 | Intron | XP_011507683.1 | |||
XM_011509382.1 | 294 | UTR 5 | XP_011507684.1 | |||
XM_011509384.2 | 294 | Intron | XP_011507686.1 | |||
XM_017000904.1 | 294 | Intron | XP_016856393.1 |