Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGTGACCACAGTGTAGAAGACA[C/G]AAATGATCTTGTTGATTTCAGGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR11L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR11L1 - olfactory receptor family 11 subfamily L member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001959.1 | 824 | Missense Mutation | TCT,TGT | S,C 275 | NP_001001959.1 |