Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAACCAGTCCCAGCGCTGTCG[C/T]GCTCCTCTACTCGGTAGGCGCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602744 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf131 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf131 - chromosome 1 open reading frame 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300830.1 | 271 | Intron | NP_001287759.1 | |||
NM_152379.3 | 271 | Intron | NP_689592.2 |
GNPAT - glyceronephosphate O-acyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316350.1 | 271 | Missense Mutation | GCG,GTG | A,V 22 | NP_001303279.1 | |
NM_014236.3 | 271 | Missense Mutation | GCG,GTG | A,V 22 | NP_055051.1 | |
XM_005273313.4 | 271 | Missense Mutation | GCG,GTG | A,V 22 | XP_005273370.1 | |
XM_011544303.2 | 271 | UTR 5 | XP_011542605.1 | |||
XM_011544304.2 | 271 | UTR 5 | XP_011542606.1 |