Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATGGCATGTATTCAGCAGGGCA[C/T]ATTATTCTTGAAGTCACAAAAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603276 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGS5 - regulator of G-protein signaling 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195303.2 | 2535 | UTR 3 | NP_001182232.1 | |||
NM_001254748.1 | 2535 | UTR 3 | NP_001241677.1 | |||
NM_001254749.1 | 2535 | UTR 3 | NP_001241678.1 | |||
NM_003617.3 | 2535 | UTR 3 | NP_003608.1 |