Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAGCAGTGTTTGAGGCACGAGA[A/G]CAGAAGGTAGGCATCTATCCTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605860 | ||||||||||||||||||||
Literature Links: |
RCAN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RCAN3 - RCAN family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251977.1 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_001238906.1 | |
NM_001251978.1 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_001238907.1 | |
NM_001251979.1 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_001238908.1 | |
NM_001251980.1 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_001238909.1 | |
NM_001251981.1 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_001238910.1 | |
NM_001251982.1 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_001238911.1 | |
NM_001251983.1 | 502 | Intron | NP_001238912.1 | |||
NM_001251984.1 | 502 | Intron | NP_001238913.1 | |||
NM_001251985.1 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_001238914.1 | |
NM_013441.3 | 502 | Silent Mutation | GAA,GAG | E,E 63 | NP_038469.1 |