Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCGTTTGTCTTCTTTAGAAGAA[A/G]TCAGGCAAACAAGTGCACACTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613173 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEGR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEGR1 - neuronal growth regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173808.2 | Intron | NP_776169.2 | ||||
XM_011541200.2 | Intron | XP_011539502.1 | ||||
XM_011541201.2 | Intron | XP_011539503.1 | ||||
XM_017000961.1 | Intron | XP_016856450.1 |