Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGGTAGCCTCATGGCTGCAACC[C/T]GTGAGATTAGCAACATTTTTAGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602191 MIM: 602675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELF3 - E74 like ETS transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114309.1 | 166 | Missense Mutation | CGT,TGT | R,C 5 | NP_001107781.1 | |
NM_004433.4 | 166 | Missense Mutation | CGT,TGT | R,C 5 | NP_004424.3 | |
XM_005244942.2 | 166 | Missense Mutation | CGT,TGT | R,C 5 | XP_005244999.1 |
MIR6740 - microRNA 6740 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNPEP - arginyl aminopeptidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |