Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTGAGCGATCTCGCGCATCAGC[C/G]GCTGGAAGGGCAACTTGCGGATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602820 MIM: 606123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIST3H3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIST3H3 - histone cluster 3, H3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003493.2 | 209 | Missense Mutation | CCG,CGG | P,R 70 | NP_003484.1 |
TRIM17 - tripartite motif containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |